Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mupirocin, 92%

Catalog No. p-7057481
Click to view available options
Quantity:
50 mg
100 mg

CAS: 12650-69-0 | C26H44O9 | 500.629 g/mol

Mupirocin is involved in inhibition of protein synthesis was reversed by isoleucine and Pseudomonic Acid was shown to inhibit IleRS (isoleucyl-tRNA synthetase) in vitro. It also demonstrates weak inhibitory action on FARSL (phenylalanyl-tRNA synthetase). Efficacy against staphylococci and streptococci including multiply resistant Staphylcoccus aureus and Staphylococcus epidermidis is reported with Pseudomonic Acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mupirocin is involved in inhibition of protein synthesis was reversed by isoleucine and Pseudomonic Acid was shown to inhibit IleRS (isoleucyl-tRNA synthetase) in vitro. It also demonstrates weak inhibitory action on FARSL (phenylalanyl-tRNA synthetase). Efficacy against staphylococci and streptococci including multiply resistant Staphylcoccus aureus and Staphylococcus epidermidis is reported with Pseudomonic Acid.

Solubility
Soluble in DMSO or methanol. Sparingly soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Typical working concentrations are in the range of 0.25µg/mL-0.39µg/mL.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12650-69-0
Molecular Formula C26H44O9
Molecular Weight (g/mol) 500.629
MDL Number MFCD01711620
InChI Key MINDHVHHQZYEEK-HBBNESRFSA-N
PubChem CID 446596
ChEBI CHEBI:7025
IUPAC Name 9-[(E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-[[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl]oxan-2-yl]-3-methylbut-2-enoyl]oxynonanoic acid
SMILES CC(C1C(O1)CC2COC(C(C2O)O)CC(=CC(=O)OCCCCCCCCC(=O)O)C)C(C)O

Specifications

Type Mupirocin
Physical Form Powder
Melting Point 77°C to 78°C
Color White
Quantity 50 mg
Flash Point >110°C (230°F)
Boiling Point 672°C
Merck Index 14,6302
Solubility Information Soluble in DMSO or methanol; Sparingly soluble in water
Formula Weight 500.62
Percent Purity 92%
Chemical Name or Material Mupirocin
Show More Show Less
RTECSNumber RA6907000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.