Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Mupirocin, 92%

Click to view available options
Quantity:
50mg
100mg
12650-69-0
C26H44O9
500.629
MFCD01711620
MINDHVHHQZYEEK-HBBNESRFSA-N
446596
CHEBI:7025
9-[(E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-[[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl]oxan-2-yl]-3-methylbut-2-enoyl]oxynonanoic acid
CC(C1C(O1)CC2COC(C(C2O)O)CC(=CC(=O)OCCCCCCCCC(=O)O)C)C(C)O
12650-69-0
C26H44O9
500.629
MFCD01711620
MINDHVHHQZYEEK-HBBNESRFSA-N
446596
CHEBI:7025
9-[(E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-[[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl]oxan-2-yl]-3-methylbut-2-enoyl]oxynonanoic acid
CC(C1C(O1)CC2COC(C(C2O)O)CC(=CC(=O)OCCCCCCCCC(=O)O)C)C(C)O
Applications
Mupirocin is involved in inhibition of protein synthesis was reversed by isoleucine and Pseudomonic Acid was shown to inhibit IleRS (isoleucyl-tRNA synthetase) in vitro. It also demonstrates weak inhibitory action on FARSL (phenylalanyl-tRNA synthetase). Efficacy against staphylococci and streptococci including multiply resistant Staphylcoccus aureus and Staphylococcus epidermidis is reported with Pseudomonic Acid.

Solubility
Soluble in DMSO or methanol. Sparingly soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Typical working concentrations are in the range of 0.25µg/mL-0.39µg/mL.
Merck Index 14,6302
Solubility Information Soluble in DMSO or methanol; Sparingly soluble in water
Formula Weight 500.62
Physical Form Powder
Percent Purity 92%
Boiling Point 672°C
Color White
Melting Point 77°C to 78°C
Quantity 100mg
Chemical Name or Material Mupirocin

RTECSNumber : RA6907000