Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mupirocin USP, Medisca™
SDP

Catalog No. 18-048-058
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g

Mupirocin USP is a white to off-white, crystalline solid.

Chemical Identifiers

CAS 12650-69-0

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystalline Solid
Color White to Off-White
Quantity 25 g
Percent Purity 920 to 1020 μg/mg
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.