Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUPP1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149975AF488
Description
MUPP1 Polyclonal antibody specifically detects MUPP1 in Human samples. It is validated for Western BlotSpecifications
MUPP1 | |
Polyclonal | |
Western Blot | |
DKFZp781P216, FLJ25909, FLJ90240, Multi-PDZ domain protein 1, multiple PDZ domain protein, MUPP1FLJ34626 | |
The immunogen for this product maps to a region between residue 1125 and 1150 of human Multiple PDZ Domain Protein using the numbering given in entry NP_003820.2 (GeneID 8777). | |
0.1 mL | |
Adaptive Immunity, Immunology | |
8777 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction