Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Murexide (Ammonium Purpurate), Reagents
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
| Melting Point | >300°C |
| Boiling Point | 1500°C |
| Packaging | Glass Bottle |
| Specific Gravity | 1 |
| pH Color Changes | Pink (Excess Calcium) to Purple (Excess EDTA) |
| Color | Purple/Red to Red |
| Quantity | 5 g |
| Physical Form | Solid |
| Chemical Name or Material | Murexide (Ammonium Purpurate) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction