Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Murexide Indicator, 0.15% (w/v) in Ethylene Glycol, Ricca Chemical
SDP

Catalog No. p-2916737
Click to view available options
Quantity:
120 mL
500 mL

APHA for Calcium Testing

Specifications

Volume (Metric) 120 mL
Packaging Amber Glass Bottle
Quantity 120 mL
Test Method APHA for Calcium
Type Murexide Indicator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.