Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Murexide Indicator, 0.2% (w/w) in Sodium Chloride, Ricca Chemical

Click to view available options
Quantity:
100 g
500 g
Specifications
Specifications
Volume (Metric) | 100 g |
Packaging | White Poly Wide Mouth Bottle |
Quantity | 100 g |
Test Method | APHA/EPA for Calcium |
Type | Murexide Indicator |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction