Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Murexide-Sodium Chloride (APHA) Water Hardness Indicator, Reagents
Click to view available options
Quantity:
100 g
250 g
500 g
Specifications
Specifications
Chemical Name or Material | Murexide-Sodium Chloride (APHA) Water Hardness Indicator |
Boiling Point | 1461°C |
CAS | 7647-14-5, 3051-09-0 |
Synonym | Ammonium Purpurate, Color Index 56085 |
Grade | Reagent |
Packaging | Glass Bottle |
Specific Gravity | 2.1 |
Color | Pink |
pH | 5 to 9 |
Quantity | 100 g |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction