Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MURF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc MURF1101AP

Description
MURF1 Polyclonal antibody specifically detects MURF1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
MURF1 | |
Polyclonal | |
Unconjugated | |
TRIM63 | |
E3 ubiquitin-protein ligase TRIM63; FLJ32380; IRF; iris ring finger protein; Murf; MURF 1; MuRF1; MuRF-1; MURF2; muscle ring finger protein 1; muscle specific ring finger protein 2; muscle-specific RING finger protein 1; OTTHUMP00000008701; RF1; ring finger protein 28; RING-type E3 ubiquitin transferase TRIM63; RNF 28; RNF28; SMRZ; striated muscle RING zinc finger protein; TRIM 63; Trim63; tripartite motif containing 63; tripartite motif containing 63, E3 ubiquitin protein ligase; tripartite motif protein 63; tripartite motif-containing 63; tripartite motif-containing protein 63 | |
Synthetic peptide taken wtihin amino acid region 270-310 on human MURF1 protein. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Q38HM4, Q91Z63, Q969Q1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
140939, 433766, 84676 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction