Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MuRF1/TRIM63 Antibody (6H6D4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | MuRF1/TRIM63 |
---|---|
Clone | 6H6D4 |
Concentration | 1 mg/mL |
Dilution | ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Description
MuRF1/TRIM63 Monoclonal specifically detects MuRF1/TRIM63 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MuRF1/TRIM63 | |
1 mg/mL | |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Zinc Finger | |
PBS with 0.05% Sodium Azide | |
84676 | |
Synthesized peptide of human MuRF1 (AA: 293-304). | |
Primary | |
40.2 kDa |
6H6D4 | |
ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 6.3.2.-, IRFMURF2, Iris RING finger protein, MuRF1, MuRF-1, muscle specific ring finger protein 2, Muscle-specific RING finger protein 1, RING finger protein 28FLJ32380, RNF28E3 ubiquitin-protein ligase TRIM63, SMRZMURF-1, Striated muscle RING zinc finger protein, tripartite motif containing 63, tripartite motif-containing 63, Tripartite motif-containing protein 63 | |
TRIM63 | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title