Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MuRF1/TRIM63 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2110610.05MG
Description
MuRF1/TRIM63 Polyclonal specifically detects MuRF1/TRIM63 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| MuRF1/TRIM63 | |
| Polyclonal | |
| Unconjugated | |
| Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide | |
| TRIM63 | |
| Synthetic peptide taken from human MURF1 protein taken from within the C-terminal domain of MURF1 corresponding to amino acid sequence. The peptide was conserved in several other species. The selected peptide was post-synthetically modified to achieve highest antigenicity before coupling to carrier protein using hetero bifunctional cross linker for immunogen preparation | |
| Primary | |
| Q969Q1 The antibody labels a strong diffuse band of MURF1of 41 kDa in PC-MURF1samples and in several muscle cell lines. The antibody also labels a diffuse band of 73kDa. The identity of this band is not known. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.50 mg/mL | |
| Western Blot 1:500, ELISA 1:500 | |
| EC 6.3.2.-, IRFMURF2, Iris RING finger protein, MuRF1, MuRF-1, muscle specific ring finger protein 2, Muscle-specific RING finger protein 1, RING finger protein 28FLJ32380, RNF28E3 ubiquitin-protein ligase TRIM63, SMRZMURF-1, Striated muscle RING zinc finger protein, tripartite motif containing 63, tripartite motif-containing 63, Tripartite motif-containing protein 63 | |
| Rabbit | |
| 0.05 mg | |
| 84676 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction