Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MURF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562826
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562826 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562826 Supplier Invitrogen™ Supplier No. PA562826
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SNDRVQGVIS QLEDTCKTIE ECCRKQKQEL CEKFDYLYGI LEERKNEMTQ VITRTQEEKL EHVRALIK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.

The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. This protein associates transiently with microtubules, myosin, and titin during muscle sarcomere assembly. It may act as a transient adaptor and plays a regulatory role in the assembly of sarcomeres. Four alternatively spliced transcript variants encoding distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MURF2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM55
Gene Accession No. Q9BYV6
Gene Alias D830041C10Rik; MURF2; MuRF-2; muscle specific ring finger 2; muscle-specific RING finger protein 2; RING finger protein 29; RNF29; Trim55; tripartite motif containing 55; tripartite motif-containing 55; tripartite motif-containing protein 55
Gene Symbols TRIM55
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MURF2. Recombinant protein control fragment (Product #RP-93811).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84675
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.