Learn More
Description
Specifications
Specifications
| Antigen | MURF3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Alias | MuRF, MuRF3, MURF-3, MURFtripartite motif-containing protein 54, Muscle-specific RING finger protein, Muscle-specific RING finger protein 3, RING finger protein 30muscle-specific RING-finger protein 3, RNF30, tripartite motif containing 54, tripartite motif-containing 54 |
| Gene Symbols | TRIM54 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
