Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MURF3 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518143
Description
This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
The protein encoded by this gene contains a RING finger motif and is highly similar to the ring finger proteins RNF28/MURF1 and RNF29/MURF2. In vitro studies demonstrated that this protein, RNF28, and RNF29 form heterodimers, which may be important for the regulation of titin kinase and microtubule-dependent signal pathways in striated muscles. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
| MURF3 | |
| Polyclonal | |
| Unconjugated | |
| TRIM54 | |
| 4930486E09Rik; 4930566I02Rik; MURF; MURF3; MuRF-3; Muscle-specific RING finger protein; Muscle-specific RING finger protein 3; muscle-specific RING-finger protein; muscle-specific RING-finger protein 3; OTTHUMP00000122598; ring finger protein 30; Rnf30; TRIM54; tripartite motif containing 54; tripartite motif-containing 54; tripartite motif-containing protein 54 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 57159 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9BYV2 | |
| TRIM54 | |
| Synthetic peptide sequence (GSAGPEEERPDGP) corresponding to the C-terminus amino acids of TRIM54 (aa 346-358). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction