Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MURF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588991
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588991 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588991 Supplier Thermo Scientific Supplier No. PA588991
Add to Cart
Add to Cart

MURF3 Polyclonal Antibody

MURF3 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen MURF3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TRIM54
Gene Accession No. Q9BYV2, Q9ERP3
Gene Alias 4930486E09Rik; 4930566I02Rik; MURF; MURF3; MuRF-3; Muscle-specific RING finger protein; Muscle-specific RING finger protein 3; muscle-specific RING-finger protein; muscle-specific RING-finger protein 3; OTTHUMP00000122598; ring finger protein 30; Rnf30; TRIM54; tripartite motif containing 54; tripartite motif-containing 54; tripartite motif-containing protein 54
Gene Symbols TRIM54
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 179-358 of human TRIM54 (NP_9127302).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57159, 58522
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.