Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Murine anti-PON1, Clone: KRJ2, ZeptoMetrix
SDP

Catalog No. 22158013 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-013 100 μg
22-158-015 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-158-013 Supplier ZeptoMetrix Supplier No. 0801014
Add to Cart
Add to Cart

Murine Monoclonal Antibody

PON1 (arylesterase/paraoxonase) is a polymorphic enzyme closely associated with high-density lipoproteins (HDL). It is located on chromosome 7 along with 2 other paraoxanase genes, PON2 and PON3. It is a 355 amino acid, 43kDa glycoprotein produced in the liver and secreted into the bloodstream. Gene polymorphisms occur in both coding and non-coding regions. Two major polymorphisms in the coding region are L55M and Q192R. Q192R polymorphisms result in three major functional phenotypes Q/Q, Q/R and RR. PON1 hydrolyses organophosphates and prevents the oxidation of low-density lipoproteins (LDL). It may inhibit the progression of atherosclerosis by inhibition of LDL oxidation. Levels of PON1 in serum vary among individuals and are decreased in chronic renal failure, liver cirrhosis, diabetes, peripheral neuropathy, and familial hypercholesterolemia.

Specifications

Antigen PON1
Applications Western Blot
Classification Monoclonal
Clone KRJ2
Conjugate Unconjugated
Formulation 100μg in PBS. No preservatives added.
Host Species Mouse
Immunogen Recombinant human PON1 produced in E. coli
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at −10°C or below. Avoid freeze/thaw cycles. Thawed material may be stored at 4°C for short-term use.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.