Learn More
Description
Specifications
Specifications
| Antigen | PON1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | KRJ2 |
| Conjugate | Unconjugated |
| Formulation | 1mg in PBS. No preservatives added. |
| Host Species | Mouse |
| Immunogen | Recombinant human PON1 produced in E. coli |
| Purification Method | Affinity Purified |
| Quantity | 1 mg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
