Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Murine Hepatitis Virus A59 nsp9 Antibody (2C6.H1), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP221671V
Description
Murine Hepatitis Virus A59 nsp9 Monoclonal specifically detects Murine Hepatitis Virus A59 nsp9 in Virus samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
nsp9 | |
Monoclonal | |
DyLight 405 | |
Non-structural protein 9, p12 | |
Mouse | |
Protein A purified | |
RUO | |
1489749 | |
Virus | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
2C6.H1 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
nsp9 | |
This Murine Hepatitis Virus A59 nsp9 Antibody (2C6.H1) was produced in mice by repeated immunizations with E.coli derived full-length Murine Hepatitis Virus A59 nsp9 protein. This protein is part of the viral replicase polyprotein. (Uniprot: P0C6V0) | |
0.1 mL | |
Primary | |
No cross reactivity occurs with SARS CoV nsp9. | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction