Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Muromonab Chimeric Recombinant Rabbit Monoclonal Antibody (OKT-3 (muromonab))
GREENER_CHOICE

Catalog No. PIMA547769
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547769 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547769 Supplier Invitrogen™ Supplier No. MA547769
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: Epitope on the epsilon-subunit within the human CD3 complex.

Murine monoclonal antibody specific to CD3 T-cell lymphocyte antigens. More specifically it is a purified murine (mouse) monoclonal antibody, directed against the CD3 (T3) receptor on the surface of human T-cells (T-lymphocytes) cultured using the murine ascites method. Muromonab is 93% monomeric immune globulin G type 2a (IgG2a). Muromonab binds to the T-cell surface glycoprotein CD3 epsilon chain. It appears to kill CD-3 positive cells by inducing Fc mediated apoptosis, antibody mediated cytotoxicity and complement-dependent cytotoxicity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Muromonab Chimeric
Applications Flow Cytometry, Immunocytochemistry
Classification Recombinant Monoclonal
Clone OKT-3 (muromonab)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Cd247
Gene Accession No. P04235, P11942, P22646, P24161
Host Species Rabbit
Immunogen Human peripheral E+ cells.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12500, 12501, 12502, 12503
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.