Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MUS81 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563743
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563743 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563743 Supplier Invitrogen™ Supplier No. PA563743
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVEEHGSVHN LSLPESTLLQ AVTNTQVIDG FFVKRTADIK ESAAYLALLT RGLQRLYQGH TLRSRPWGTP GNPESGAMTS PNPLCSLLTF SDFNAGAIKN KAQSVREVFA RQLMQVRGVS GEKAAALVDR YSTPA Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

Interacts with EME1 and EME2 to form a DNA structure-specific endonuclease with substrate preference for branched DNA structures with a 5'-end at the branch nick. Typical substrates include 3'-flap structures, replication forks and nicked Holliday junctions. May be required in mitosis for the processing of stalled or collapsed replication forks.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUS81
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MUS81
Gene Accession No. Q96NY9
Gene Alias 1200008A18Rik; AI182501; AW045863; crossover junction endonuclease MUS81; FLJ21012; FLJ44872 MUS81; MUS81; MUS81 endonuclease homolog; MUS81 endonuclease homolog (yeast); MUS81 structure-specific endonuclease subunit; SLX3; SLX3 structure-specific endonuclease subunit homolog
Gene Symbols MUS81
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MUS81. Recombinant protein control fragment (Product # RP-89866).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80198
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.