Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MUS81 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143846
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143846 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143846 Supplier Invitrogen™ Supplier No. PA5143846
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue. Flow: 293T cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Interacts with EME1 and EME2 to form a DNA structure-specific endonuclease with substrate preference for branched DNA structures with a 5'-end at the branch nick. Typical substrates include 3'-flap structures, replication forks and nicked Holliday junctions. May be required in mitosis for the processing of stalled or collapsed replication forks.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUS81
Applications ELISA, Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene MUS81
Gene Accession No. Q4KM32, Q96NY9
Gene Alias 1200008A18Rik; AI182501; AW045863; crossover junction endonuclease MUS81; FLJ21012; FLJ44872 MUS81; MUS81; MUS81 endonuclease homolog; MUS81 endonuclease homolog (yeast); MUS81 structure-specific endonuclease subunit; SLX3; SLX3 structure-specific endonuclease subunit homolog
Gene Symbols MUS81
Host Species Rabbit
Immunogen E.coli-derived human MUS81 recombinant protein (Position: R6-C545).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293678, 80198
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.