Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Musashi-1 Antibody (OTI1G8), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272828MFV500
Description
Musashi-1 Monoclonal antibody specifically detects Musashi-1 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Flow CytometrySpecifications
Musashi-1 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Primate, Monkey | |
Purified |
Flow Cytometry | |
OTI1G8 | |
Flow Cytometry | |
Musashi (Drosophila) homolog 1, musashi homolog 1 (Drosophila), Musashi-1, RNA-binding protein Musashi homolog 1 | |
Full length human recombinant protein of human MSI1 (NP_002433) produced in HEK293T cell. | |
0.1 mL | |
Cancer, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells | |
4440 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction