Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Musashi-2 Antibody (960121) [DyLight 350], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3255D
Description
Musashi-2 Monoclonal antibody specifically detects Musashi-2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
Musashi-2 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
960121 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
FLJ36569, MGC3245, MSI2H, musashi homolog 2 (Drosophila), musashi-2, RNA-binding protein Musashi homolog 2 | |
E. coli-derived recombinant human Musashi-2, Met1-His328, Accession # Q96DH6 | |
0.1 mL | |
Neuronal Cell Markers | |
124540 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction