Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Musashi-2 Antibody (OTI2F10), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272836PE
Description
Musashi-2 Monoclonal antibody specifically detects Musashi-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
Musashi-2 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
OTI2F10 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
FLJ36569, MGC3245, MSI2H, musashi homolog 2 (Drosophila), musashi-2, RNA-binding protein Musashi homolog 2 | |
Full length human recombinant protein of human MSI2(NP_620412) produced in HEK293T cell. | |
0.1 mL | |
Neuronal Cell Markers | |
124540 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction