Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Muscarinic Acetylcholine Receptor M1/CHRM1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB10041399
Description
Muscarinic Acetylcholine Receptor M1/CHRM1 Polyclonal specifically detects Muscarinic Acetylcholine Receptor M1/CHRM1 in Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA.Specifications
Muscarinic Acetylcholine Receptor M1/CHRM1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
cholinergic receptor, muscarinic 1, HM1, M1, M1R, MGC30125, muscarinic acetylcholine receptor M1 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Cross reacts with Human and Rat. Expected from sequence similarity: Mouse and Dog. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:64000 | |
NP_000729.2 | |
CHRM1 | |
Peptide with sequence C-DKRRWRKIPKRPGSVH corresponding to C-Terminus according to NP_000729.2. | |
0.1 mg | |
Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Transcription Factors and Regulators | |
1128 | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction