Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Muscarinic Acetylcholine Receptor M2/CHRM2 Antibody (SR1913), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Muscarinic Acetylcholine Receptor M2/CHRM2 |
---|---|
Clone | SR1913 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
Classification | Monoclonal |
Description
Muscarinic Acetylcholine Receptor M2/CHRM2 Monoclonal antibody specifically detects Muscarinic Acetylcholine Receptor M2/CHRM2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
Muscarinic Acetylcholine Receptor M2/CHRM2 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
7TM receptor, cholinergic receptor, muscarinic 2, FLJ43243, HM2, MGC120006, MGC120007, muscarinic acetylcholine receptor M2, muscarinic M2 receptor | |
A synthesized peptide derived from human Muscarinic Acetylcholine Receptor M2/CHRM2 (Uniprot #: P08172) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1913 | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
GPCR, Neuroscience | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
1129 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title