Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Muscarinic Acetylcholine Receptor M2/CHRM2 Antibody (585713), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6377Y
This item is not returnable.
View return policy
Description
Muscarinic Acetylcholine Receptor M2/CHRM2 Monoclonal antibody specifically detects Muscarinic Acetylcholine Receptor M2/CHRM2 in Human samples. It is validated for Flow Cytometry, ImmunohistochemistrySpecifications
Muscarinic Acetylcholine Receptor M2/CHRM2 | |
Monoclonal | |
DyLight 680 | |
7TM receptor, cholinergic receptor, muscarinic 2, FLJ43243, HM2, MGC120006, MGC120007, muscarinic acetylcholine receptor M2, muscarinic M2 receptor | |
NS0 mouse myeloma cell line transfected with human CHRM2, Accession # P08172 | |
0.1 mL | |
GPCR, Neuroscience | |
1129 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, Immunohistochemistry | |
585713 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction