Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Muscarinic Acetylcholine Receptor M4/CHRM4 Antibody (18C7.2), Novus Biologicals™
SDP

Catalog No. p-7200121 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019435 0.025 mL
NBP229705 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019435 Supplier Novus Biologicals Supplier No. NBP2297050.025ML
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Muscarinic Acetylcholine Receptor M4/CHRM4 Monoclonal specifically detects Muscarinic Acetylcholine Receptor M4/CHRM4 in Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen Muscarinic Acetylcholine Receptor M4/CHRM4
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 18C7.2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500
Formulation Ascites with 20mM Phosphate and 0.25M NaCl with 0.1% Sodium Azide
Gene Accession No. P08173
Gene Alias cholinergic receptor, muscarinic 4, HM4, M4R, muscarinic acetylcholine receptor M4
Gene Symbols CHRM4
Host Species Mouse
Immunogen m4 receptor i3 loop (human), fused to GST.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 1132
Test Specificity m4 muscarinic acetylcholine receptor. No reactivity with the other subtypes.
Target Species Monkey
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.