Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Muscle Actin Mouse anti-Human, Clone: MSA06, Abnova™

Catalog No. 89934635 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-635 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-635 Supplier Abnova Corporation Supplier No. MAB12630

Mouse monoclonal antibody raised against human Muscle Actin.

Specifications

Antigen Muscle Actin
Applications Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone MSA06
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human Muscle Actin.
Dilution Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide).
Host Species Mouse
Immunogen Human Muscle Actin
Purification Method Protein G purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.