Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cenmed™ Muscle Biopsy Kit with Glutaraldehyde 3% in 0.1 Phosphate Buffer

Click to view available options
Quantity:
100 Kits/Pk.
Description
- 3 oz. Formalin Buffered to 10%, providing reliable preservation in a convenient half-filled container
- Glutaraldehyde 3% in 0.1 Phosphate Buffer ensures optimal tissue fixation, contained in a convenient 5.2 mL vial
- Safely store and transport your specimens with clear-orange biohazard specimen bags, measuring 6 x 9 in. and equipped with indicators for added safety assurance
- Streamline your sample processing with 15 mL polypropylene centrifuge tubes, measuring 17 x 118 mm, provided in racks for organized and sterile handling
- For larger sample volumes, 50 mL sterile centrifuge tubes, complete with trays, offer dependable containment for diverse laboratory applications
- Specialized laboratory products enhance efficiency and accuracy in your experiments, setting a new standard for excellence in laboratory practices
Specifications
Specifications
Product Type | Specimen Collection Kit |
Format | Tube |
Media Type | Molecular Transport Media |
For Use With (Application) | Collection, Transport, and Preservation of Biological Specimens |
Quantity | 100 Kits/Pk. |
Material | Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction