Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Muscle Phosphofructokinase/PFKM/PFK-1 Rabbit anti-Human, Mouse, Rat, Clone: 4Z7X1, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Muscle Phosphofructokinase/PFKM/PFK-1 |
---|---|
Clone | 4Z7X1 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Muscle Phosphofructokinase/PFKM/PFK-1 Monoclonal antibody specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Muscle Phosphofructokinase/PFKM/PFK-1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5213 | |
IgG | |
Affinity purified |
4Z7X1 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, phosphofructokinase-M, phosphohexokinase | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human Muscle Phosphofructokinase/PFKM/PFK-1 (P08237). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title