Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Muscleblind-like 1 Antibody (1H1), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250062AF594
Description
Muscleblind-like 1 Monoclonal specifically detects Muscleblind-like 1 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Muscleblind-like 1 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
MBNL1 | |
Full-length recombinant human muscleblind-like 1 protein expressed in and purified from E. coli. [UniProt# Q9NR56] | |
0.1 mL | |
Primary | |
Human, Mouse (Negative), Rat (Negative) | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
1H1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
DKFZp686P06174, EXP40, EXP42, EXPKIAA0428EXP35, MBNL, muscleblind (Drosophila)-like, muscleblind-like (Drosophila), muscleblind-like protein 1, Triplet-expansion RNA-binding protein | |
Mouse | |
Affinity Purified | |
Chromatin Research, DNA replication Transcription Translation and Splicing | |
4154.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction