Learn More
Eisco™ Muscovite Mica Mineral Specimen
SCI_ED
Catalog No. S24704 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24704 1 Ea. 2 to 3 cm
S26598 10/Pk. 2 to 3 cm
S24705 12/Pk. 2 to 3 cm
S26657 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24704 Supplier Eisco™ Supplier No. ESNG0022
Add to Cart
Add to Cart
Muscovite Mica Mineral Specimen forms in thin sheets that are individually weak, but comprehensively strong.
  • Unique thin layered structure
  • For use in classroom identification labs and exercises
  • For use in study of variable hardness and cleavage
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Pair with biotite mica to demonstrate effect of slight variations in mineral's chemistry
Specifications
Product Type Mineral
For Use With (Application) Study of variable hardness and cleavage
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Specimen Type Muscovite Mica