Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MuSK Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1205619
Description
MuSK Polyclonal specifically detects MuSK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
MuSK | |
Polyclonal | |
Unconjugated | |
PBS, 1.0 mg/ml BSA and 30% Glycerol with 0.05% Sodium Azide | |
MUSK | |
Recombinant protein containing residues 210-304 from the extracellular domain of rat MuSK | |
200 μg | |
Protein Kinase, Signal Transduction | |
4593 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1:500 - 1:1000, Immunohistochemistry 1:20 - 1:100, Immunocytochemistry/Immunofluorescence 1:20 - 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20 - 1:100 | |
EC 2.7.10, EC 2.7.10.1, MGC126323, MGC126324, muscle, skeletal, receptor tyrosine kinase, MuSK, skeletal receptor tyrosine-protein kinase | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Detects MUSK from mouse and rat tissue samples as well as COS7 cells transfected with rat MUSK. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction