Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MUSK Monoclonal Antibody (1429CT456.173.44)
GREENER_CHOICE

Catalog No. PIMA537664
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537664 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537664 Supplier Invitrogen™ Supplier No. MA537664
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MUSK Monoclonal Antibody for Western Blot, IHC (P)

MUSK plays an important and selective role in many aspects of skeletal muscle development and function through the initial step of postsynaptic differentiation and formation of end-plate bands in cooperation with Rapsyn. Communication between cells is often mediated by receptors on the surface of the target cell that are triggered by specific ligands released by other cells. Receptor tyrosine kinase activity is strictly regulated by the binding of the appropriate ligand to the extracellular active site of the receptor. MuSK has been shown to be part of the agrin signaling complex and plays an important role in the development and architectural maintenance of the neuromuscular junction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUSK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1429CT456.173.44
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MUSK
Gene Accession No. Q61006, Q62838
Gene Alias CMS9; FADS; Mdk4; MGC126323; MGC126324; Mlk; muscle associated receptor tyrosine kinase; muscle localized kinase 2; muscle specific kinase (neural fold/somite kinase 1); muscle, skeletal receptor tyrosine protein kinase; Muscle, skeletal receptor tyrosine-protein kinase; muscle, skeletal, receptor tyrosine kinase; muscle-specific kinase receptor; muscle-specific protein kinase secretory isoform; muscle-specific tyrosine kinase receptor MuSK; muscle-specific tyrosine protein kinase receptor; Muscle-specific tyrosine-protein kinase receptor; MuSK; Nsk1; Nsk2; Nsk3; RP23-170A3.1
Gene Symbols MUSK
Host Species Mouse
Immunogen recombinant protein from mouse Musk.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18198, 81725
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.