Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Muskelin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554611
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554611 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554611 Supplier Invitrogen™ Supplier No. PA554611
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QAAKDNPTKS LQEEEPCPRF AHQLVYDELH KVHYLFGGNP GKSCSPKMRL DDFWSLKLCR PSKDYLLRHC KYLIRKHRFE EKAQVDPLSA LKYLQNDLYI TVDHSDP Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

Muskelin is an intracellular protein that acts as a mediator of cell spreading and cytoskeletal responses to the extracellular matrix component thrombospondin I.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Muskelin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MKLN1
Gene Accession No. Q9UL63
Gene Alias A130067F06Rik; AU015903; BB154892; cytoplasmic mediator; fc60a07; LOW QUALITY PROTEIN: muskelin; Mkln1; mkln1 protein; Msk; Muskelin; muskelin 1; muskelin 1, intracellular mediator containing kelch motifs; TWA2; wu:fc60a07
Gene Symbols MKLN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MNK1. Recombinant protein control fragment (Product #RP-92714).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4289
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.