Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Muskelin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588356
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588356 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588356 Supplier Invitrogen™ Supplier No. PA588356
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QTWKLLREDS CNAGPEDIQS RIGHCMLFHS KNRCLYVFGG QRSKTYLNDF FSYDVDSDHV DIISDGTKKD SGMVPMTGFT QRATIDPELN EIHVLSGLSK DKEKREENVR NSFWIYDIVR NSWSCVYKND QAAKDNPTKS LQEEEPCPRF AHQLVYDELH KVHYLFGGNP GKSCSPKMRL DDFWSLKLCR PSKDYLLRHC KYLIRKHRFE EKAQVDPLSA LKYLQNDLYI TVDHSDPEET KEFQLLASAL FKSGSDFTAL GFSDVDHTYA QRTQLFDTLV NFFPDSMTPP KGNLVDLITL; Positive Samples: Mouse spleen; Cellular Location: Cell projection, Cytoplasm, cell cortex, ruffle.

Muskelin is an intracellular protein that acts as a mediator of cell spreading and cytoskeletal responses to the extracellular matrix component thrombospondin I.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Muskelin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MKLN1
Gene Accession No. O89050
Gene Alias A130067F06Rik; AU015903; BB154892; cytoplasmic mediator; fc60a07; LOW QUALITY PROTEIN: muskelin; Mkln1; mkln1 protein; Msk; Muskelin; muskelin 1; muskelin 1, intracellular mediator containing kelch motifs; TWA2; wu:fc60a07
Gene Symbols MKLN1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 436-735 of human MKLN1 (NP_0373872).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27418
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.