Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Muskelin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599034
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599034 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599034 Supplier Invitrogen™ Supplier No. PA599034
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Muskelin Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Muskelin is an intracellular protein that acts as a mediator of cell spreading and cytoskeletal responses to the extracellular matrix component thrombospondin I.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Muskelin
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MKLN1
Gene Accession No. Q99PV3, Q9UL63
Gene Alias A130067F06Rik; AU015903; BB154892; cytoplasmic mediator; fc60a07; LOW QUALITY PROTEIN: muskelin; Mkln1; mkln1 protein; Msk; Muskelin; muskelin 1; muskelin 1, intracellular mediator containing kelch motifs; TWA2; wu:fc60a07
Gene Symbols MKLN1
Host Species Rabbit
Immunogen Recombinant Human Muskelin protein (488-614AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4289, 83536
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.