Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Muskelin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501729682 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9682 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9682 Supplier Proteintech Group Inc Supplier No. 147351AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAP kinase-interacting serine/threonine kinases (Mnk) function in signal transduction pathways through phosphorylation of eIF4E and are directly phosphorylated by ERK or p38 MAP kinases. In human and mouse there are two Mnk genes. Mnk1 is activated by phosphorylation at Thr197 and Thr202. Mnk1 and Mnk2 are essential for the phosphorylation of eIF4E at Ser209 however they differ markedly in their activity and regulatory patterns. Mnk1 is responsible for inducible phosphorylation of eIF4E while Mnk2 is responsible for basal levels of phosphorylation. Mnk1 has been implicated in signaling in response to oxidative stress by increasing eIF4E phosphorylation upon H2O2 treatment in mice. Oxidative stress induced Mnk1 activity has also been linked to hyperproliferative diseases through the over-activation of eIF4E. Reactivity with Mnk2 has not been tested.

Specifications

Antigen Muskelin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MKLN1
Gene Accession No. O89050, Q99PV3, Q9UL63
Gene Alias MKLN1, Muskelin, TWA2
Gene Symbols MKLN1
Host Species Rabbit
Immunogen Muskelin Fusion Protein Ag6147
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27418, 4289, 83536
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.