Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUT Mouse anti-Human, Clone: OTI2A8, TrueMAB™, Origene

Mouse Monoclonal Antibody
Supplier: OriGene TA506883S

This item is not returnable.
View return policy
Description
MUT Monoclonal antibody specifically detects MUT in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
MUT | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P22033 | |
Mouse | |
Affinity chromatography | |
RUO | |
4594 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
OTI2A8 | |
Unconjugated | |
MMUT | |
D230010K02Rik; Mcm; methylmalonyl CoA mutase; methylmalonyl Coenzyme A mutase; methylmalonyl-CoA isomerase; methylmalonyl-CoA mutase; methylmalonyl-CoA mutase, mitochondrial; methylmalonyl-Coenzyme A mutase; MMUT; MUT; mutant methylmalonyl CoA mutase; truncated methylmalonyl CoA mutase | |
Full length human recombinant protein of human MUT produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction