Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mutant HTT Mouse anti-Human,Mouse, Unconjugated, Clone: PHP4, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $457.74
Specifications
Antigen | mutant HTT |
---|---|
Clone | PHP4 |
Applications | Dot Blot, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABN25402MI
|
MilliporeSigma
MABN254025UL |
25 μL |
Each for $182.31
|
|
|||||
MABN25401MI
|
MilliporeSigma
MABN2540100UL |
100 μL |
Each for $457.74
|
|
|||||
Description
mutant HTT Monoclonal antibody specifically detects mutant HTT in Human,Mouse samples. It is validated for Dot Blot, Electron Microscopy, Immunohistochemistry (Paraffin), Inhibits Activity/Function, Western Blotting.Specifications
mutant HTT | |
Dot Blot, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
Huntingtin;Huntington disease protein;HD protein | |
GST-tagged recombinant exon1 of wild-type human huntingtin HTTx1 (20Q) protein. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. | |
347.60 kDa calculated. |
PHP4 | |
Monoclonal | |
Purified | |
Neuroscience | |
NP_002102.4 | |
HTT, HD, IT15 | |
IgG1 | |
Protein G Purified | |
Clone PHP4 is a mouse monoclonal antibody that detects Huntington protein. It targets an epitope within polyQ-polyP junction (QQQQPPPP). It predominantly reacts with soluble mutant HTT. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title