Learn More
Invitrogen™ MUTED Polyclonal Antibody

Description
This antibody is predicted to react with porcine based on sequence homology.
Specifications
Specifications
| Antigen | MUTED |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | BLOC1S5 |
| Gene Accession No. | Q8R015, Q8TDH9 |
| Gene Alias | 1810074A19Rik; biogenesis of lysosomal organelles complex 1 subunit 5; biogenesis of lysosomal organelles complex-1, subunit 5, muted; biogenesis of lysosome-related organelles complex 1 subunit 5; biogenesis of organelles complex-1, subunit 5, muted; BLOC-1 subunit 5; Bloc1s5; BLOS5; MU; Muted; muted homolog; protein Muted homolog |
| Gene Symbols | BLOC1S5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.