Learn More
Invitrogen™ MUTYH Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, NIH-3T3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | MUTYH |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.16 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | MUTYH |
| Gene Accession No. | Q99P21, Q9UIF7 |
| Gene Alias | 5730495A01Rik; A/G-specific adenine DNA glycosylase; adenine DNA glycosylase; adenine-DNA glycosylase; hMYH; MGC4416; mMYH; mutY DNA glycosylase; mutY homolog; mutY homolog (E. coli); mutY homolog alpha; Mutyh; Mutyha; Mutyhalpha; Mutyhb; Mutyhbeta; Mutyhc; mutY-like protein; Myh; OTTHUMP00000009098; OTTHUMP00000009102; rMYH; RP4-534D1.2; the full-length form type A, 5' region |
| Gene Symbols | MUTYH |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.