Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MUTYH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110751
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110751 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110751 Supplier Invitrogen™ Supplier No. PA5110751
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PDVEECAPNT GQCHLCLPPS EPWDQTLGVV NFPRKASRKP PREESSATCV LEQPGALGAQ ILLVQRPNSG LLAGLWEFPS VTWEPSEQLQ RKALLQELQR WA Highest antigen sequence indentity to the following orthologs: Mouse - 77%, Rat - 78%.

MYH (mutY homolog (E. coli )) is a DNA glycosylase mismatch repair enzyme that in conjunction with mutM (OGG1), cleaves adenine residues paired with either oxidized (8-hydroxyguanines) or non-modified guanines in order to correct A/G and A/C mismatches. Repair of most modified and mispaired bases in the genome is initiated by DNA glycosylases, which bind and cleave N-glycosyl bonds to initiate base excision repair. MYH is crucial for the avoidance of mutations resulting from oxidative DNA damage. Multiple N-terminal splice variants of MYH exist in mammalian cells. Increasing levels of MYH in A549 cells exposed to oxygen and infrared radiation leads to improvements in cell survival. Biallelic MYH germ-line mutations predispose humans to colorectal adenomas and carcinomas. MYH is abundant in neurons where mitochondrial genomes exposed to reactive oxygen species (ROS) that damage DNA must maintain integrity over the entire mammalian life span.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUTYH
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MUTYH
Gene Accession No. Q9UIF7
Gene Alias 5730495A01Rik; A/G-specific adenine DNA glycosylase; adenine DNA glycosylase; adenine-DNA glycosylase; hMYH; MGC4416; mMYH; mutY DNA glycosylase; mutY homolog; mutY homolog (E. coli); mutY homolog alpha; Mutyh; Mutyha; Mutyhalpha; Mutyhb; Mutyhbeta; Mutyhc; mutY-like protein; Myh; OTTHUMP00000009098; OTTHUMP00000009102; rMYH; RP4-534D1.2; the full-length form type A, 5' region
Gene Symbols MUTYH
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human MUTYH. Recombinant protein control fragment (Product #RP-89477).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4595
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.