Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MUTYH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595692
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595692 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595692 Supplier Invitrogen™ Supplier No. PA595692
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTPLVSRLSR LWAIMRKPRA AVGSGHRKQA ASQEGRQKHA KNNSQAKPSA CDGLARQPEE VVLQASVSSY HLFRDVAEVT AFRGSLLSWY DQEKRDLPWR RRAEDEMDLD RRAYAVWVSE VMLQQTQVAT VINYYTGWMQ KWPTLQDLAS ASLEEVNQLW AGLGYYSRGR RLQEGARKVV EELGGHMPRT AETLQQLLPG VGRYTAGAIA SIAFGQATGV VDGNVARVLC RVRAIGADPS STLVSQQLWG LAQQLVDPAR PGDFN.

MYH (mutY homolog (E. coli )) is a DNA glycosylase mismatch repair enzyme that in conjunction with mutM (OGG1), cleaves adenine residues paired with either oxidized (8-hydroxyguanines) or non-modified guanines in order to correct A/G and A/C mismatches. Repair of most modified and mispaired bases in the genome is initiated by DNA glycosylases, which bind and cleave N-glycosyl bonds to initiate base excision repair. MYH is crucial for the avoidance of mutations resulting from oxidative DNA damage. Multiple N-terminal splice variants of MYH exist in mammalian cells. Increasing levels of MYH in A549 cells exposed to oxygen and infrared radiation leads to improvements in cell survival. Biallelic MYH germ-line mutations predispose humans to colorectal adenomas and carcinomas. MYH is abundant in neurons where mitochondrial genomes exposed to reactive oxygen species (ROS) that damage DNA must maintain integrity over the entire mammalian life span.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUTYH
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MUTYH
Gene Accession No. Q8R5G2, Q9UIF7
Gene Alias 5730495A01Rik; A/G-specific adenine DNA glycosylase; adenine DNA glycosylase; adenine-DNA glycosylase; hMYH; MGC4416; mMYH; mutY DNA glycosylase; mutY homolog; mutY homolog (E. coli); mutY homolog alpha; Mutyh; Mutyha; Mutyhalpha; Mutyhb; Mutyhbeta; Mutyhc; mutY-like protein; Myh; OTTHUMP00000009098; OTTHUMP00000009102; rMYH; RP4-534D1.2; the full-length form type A, 5' region
Gene Symbols MUTYH
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-265 of human MUTYH (NP_0010416361).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170841, 4595
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.