Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MVB12A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559924
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559924 Supplier Invitrogen™ Supplier No. PA559924
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ISCTVEGAPA SFGKSFAQKS GYFLCLSSLG SLENPQENVV ADIQIVVDKS PLPLGFSPVC DPMDSKASVS KKKRMCVKLL PL Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 74%.

FAM125A, also named as MVB12A and CFBP, is a component of the ESCRT-I complex. FAM125A is required for the sorting of endocytic ubiquitinated cargos into multivesicular bodies. FAM125A has three isoforms with MW 27-28 kDa and 25 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MVB12A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MVB12A
Gene Accession No. Q96EY5
Gene Alias CFBP; CIN85/CD2AP family binding protein; CIN85/CD2AP family-binding protein; ESCRT-I complex subunit MVB12A; Fam125a; family with sequence similarity 125, member A; multivesicular body subunit 12A; Mvb12a; Protein FAM125A
Gene Symbols MVB12A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MVB12A. Recombinant protein control fragment (Product #RP-99120).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93343
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.