Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MVB12A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590833
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590833 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590833 Supplier Invitrogen™ Supplier No. PA590833
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDPVPGTDSA PLAGLAWSSA SAPPPRGFSA ISCTVEGAPA SFGKSFAQKS GYFLCLSSLG SLENPQENVV ADIQIVVDKS PLPLGFSPVC DPMDSKASVS KKKRMCVKLL PLGATDTAVF DVRLSGKTKT VPGYLRIGDM GGFAIWCKKA KAPRPVPKPR GLSRDMQGLS LDAASQPSKG GLLERTASRL GSRASTLRRN DSIYEASSLY GISAMDGVPF TLHPRFEGKS CSPLAFSAFG DLTIKSLADI EEEYNYGFVV EKTAAARLPP SVS; Positive Samples: Rat pancreas; Cellular Location: Cytoplasm, Endosome, Late endosome membrane, Nucleus, Peripheral membrane protein, centrosome, cytoskeleton, microtubule organizing center.

FAM125A, also named as MVB12A and CFBP, is a component of the ESCRT-I complex. FAM125A is required for the sorting of endocytic ubiquitinated cargos into multivesicular bodies. FAM125A has three isoforms with MW 27-28 kDa and 25 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MVB12A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MVB12A
Gene Accession No. Q6P777
Gene Alias CFBP; CIN85/CD2AP family binding protein; CIN85/CD2AP family-binding protein; ESCRT-I complex subunit MVB12A; Fam125a; family with sequence similarity 125, member A; multivesicular body subunit 12A; Mvb12a; Protein FAM125A
Gene Symbols MVB12A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-273 of human MVB12A (NP_6124101).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 290635
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.