Learn More
Invitrogen™ MVB12A Polyclonal Antibody

Description
Immunogen sequence: LGATDTAVFD VRLSGKTKTV PGYLRIGDMG GFAIWCKKAK APRPVPKPRG LSRDMQGLSL DAASQPSKGG LLERTASRLG SRASTL Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 73%.
Specifications
Specifications
| Antigen | MVB12A |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MVB12A |
| Gene Accession No. | Q96EY5 |
| Gene Alias | CFBP; CIN85/CD2AP family binding protein; CIN85/CD2AP family-binding protein; ESCRT-I complex subunit MVB12A; Fam125a; family with sequence similarity 125, member A; multivesicular body subunit 12A; Mvb12a; Protein FAM125A |
| Gene Symbols | MVB12A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.