Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MVD Recombinant Rabbit Monoclonal Antibody (7G4B9)
GREENER_CHOICE

Catalog No. PIMA542627
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542627 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542627 Supplier Invitrogen™ Supplier No. MA542627
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: HepG2, Rat brain. Immunogen sequence: PSFAQLTMKD SNQFHATCLD TFPPISYLNA ISWRIIHLVH RFNAHHGDTK VAYTFDAGPN AVIFTLDDTV AEFVAAVWHG FPPGSNGDTF LKGLQVRPAP L.

The enzyme mevalonate pyrophosphate decarboxylase catalyzes the conversion of mevalonate pyrophosphate into isopentenyl pyrophosphate in one of the early steps in cholesterol biosynthesis. It decarboxylates and dehydrates its substrate while hydrolyzing ATP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MVD
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 7G4B9
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Mvd
Gene Accession No. P53602, Q62967
Gene Alias C78718; diphosphomevalonate decarboxylase; FP17780; MDDase; mevalonate (diphospho) decarboxylase; Mevalonate (diphospho)decarboxylase; mevalonate diphosphate decarboxylase; mevalonate pyrophosphate decarboxylase; Mpd; MVD; Mvpd; POROK7
Gene Symbols Mvd
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 250-350 of human MVD (P53602).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4597, 81726
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.