Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MVK Antibody (OTI 1D7) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272837
Description
MVK Monoclonal specifically detects MVK in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
MVK | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
EC 2.7.1.36, LH receptor mRNA-binding protein, LRBP, mevalonate kinase, mevalonate kinase (mevalonic aciduria), mevalonate kinase 1, MKFLJ96772, MVLK | |
Mouse | |
42.3 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI 1D7 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
MVK | |
Full length human recombinant protein of human MVK (NP_000422) produced in HEK293T cell. | |
Affinity Purified | |
metabolism | |
4598 | |
Human | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction